Learn More
Analytical Sales & Services 48-Position Silicone/PTFE Cap Mat for HPLC Vials in Waters and Aquity Autosampler Tray, Clear, Pre-slit
Catalog No. 5000805910
Encompass
48-position Silicone/PTFE Cap Mats Clear, Pre-Slit, for 2mL, 9mm, 12 x 32mm HPLC Vials in the Waters Aquity Autosampler Tray\n\n• Pre-slit\n• Fits 2mL, 9mm, 12 x 32mm HPLC vials\n• Material: silicone/PTFE\n• Color: clear\n• Efficiently cap 48 HPLC vials at once!\n• Great for dissolution assays\n• Made in the USA\n\nQty/Pkg: 5
Catalog No. 50-008-05910 Supplier Analytical Sales & Services Supplier No. 99975
48-position Silicone/PTFE Cap Mats Clear, Pre-Slit, for 2mL, 9mm, 12 x 32mm HPLC Vials in the Waters Aquity Autosampler Tray\n\n• Pre-slit\n• Fits 2mL, 9mm, 12 x 32mm HPLC vials\n• Material: silicone/PTFE\n• Color: clear\n• Efficiently cap 48 HPLC vials at once!\n• Great for dissolution assays\n• Made in the USA\n\nQty/Pkg: 5