Learn More
Viva Products NoEndoTM Endotoxin Removal Spin Columns, High Capacity
SDP
Supplier:  Viva Products GENNOE48HC
Encompass
Proteus NoEndoTM Spin Column Kit; For fast affinity based removal of endotoxins from antibodies and other protein samples; 20 ml capacity; High Capacity resin can remove over 500,000 EU of endotoxin; 48/PK
Catalog No. NC0941508