Learn More
CELLTREAT Scientific Products 48 Well Tissue Culture Plate, Sterile
SDP
Catalog No. 50607510
Encompass_Preferred
CELLTREAT Scientific Products 48 Well Tissue Culture Treated Plate with Lid. Tissue Culture Treated surface improves cell adhesion. Plates are labeled with alpha-numeric markings for easy identification of individual wells. Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination. Lids have a stacking feature for added stability when handling tall stacks of plates. Flat bottom with cell growth area of 0.84cm¿. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Individually packed sterile, 50/case.
Catalog No. 50-607-510 Supplier CELLTREAT Scientific Products Supplier No. 229147
CELLTREAT Scientific Products 48 Well Tissue Culture Treated Plate with Lid. Tissue Culture Treated surface improves cell adhesion. Plates are labeled with alpha-numeric markings for easy identification of individual wells. Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination. Lids have a stacking feature for added stability when handling tall stacks of plates. Flat bottom with cell growth area of 0.84cm¿. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Individually packed sterile, 50/case.