Learn More
Arcticwhite LLC 48 Rectangle Well, 5.0 ML, Polypropylene, Pyramid Bottom, 48 Wells, ANSI/SLAS Standard Base and Footprint, 5.0 ML/well, 25 Microplates per Case
SDP
Catalog No. NC9948863
Encompass
48 Rectangle Well, 5.0 ML/Well, Polypropylene, Pyramid Bottom, 48 Wells, ANSI/SLAS Standard Base and Footprint, This 48-well microplate is used for mixing, sample recovery, automated liquid handling, storage, and downstream processing in research and diagnostics laboratories. Format - 48 wells, ANSI/SLAS standard base and footprint. Well shape - Rectangle. Bottom - Pyramid. Well volume - 5.0 mL/well. Material - Polypropylene. Supplied by ArcticWhite as 25 microplates per case.
Catalog No. NC9948863 Supplier Arcticwhite LLC Supplier No. AWLS360002
48 Rectangle Well, 5.0 ML/Well, Polypropylene, Pyramid Bottom, 48 Wells, ANSI/SLAS Standard Base and Footprint, This 48-well microplate is used for mixing, sample recovery, automated liquid handling, storage, and downstream processing in research and diagnostics laboratories. Format - 48 wells, ANSI/SLAS standard base and footprint. Well shape - Rectangle. Bottom - Pyramid. Well volume - 5.0 mL/well. Material - Polypropylene. Supplied by ArcticWhite as 25 microplates per case.