Learn More
Analytical Sales & Services 48-Well Storage/Reaction Microplate, 7.5mL
Catalog No. 5000804520
Encompass
48-well Polypropylene Storage/reaction Microplate, 7.5mL/rectangle well, pyramid bottoms, 68mm height 8 rows x 6 columns
Catalog No. 50-008-04520 Supplier Analytical Sales & Services Supplier No. 48687
48-well Polypropylene Storage/reaction Microplate, 7.5mL/rectangle well, pyramid bottoms, 68mm height 8 rows x 6 columns