Learn More
USA Scientific Inc TempPlate non-skirted 48-well PCR plate, chimney, natural
Catalog No. NC9799385
Encompass_Preferred
TempPlate non-skirted polypropylene 0.2 mL 48-well PCR plate, chimney wells, standard depth, natural. 2/sleeve, 20/box.
Catalog No. NC9799385 Supplier USA Scientific Inc Supplier No. 14020480
TempPlate non-skirted polypropylene 0.2 mL 48-well PCR plate, chimney wells, standard depth, natural. 2/sleeve, 20/box.