Learn More
Bellco Glass, Inc. 48 Well Tissue Culture Plate
SDP
Catalog No. 502330603
Encompass_Preferred
Tissue Culture Treated surface improves cell adhesionPlates are labeled with alpha-numeric markings for easy identification of individual wellsEach well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contaminationLids have a stacking feature for added stability when handling tall stacks of plates Pack DescriptionIndividual, 50/PackCase Quantity100SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPolystyreneTissue Culture TreatedYesBottom ShapeFlatCell Growth Area0.84cm²RNase/DNase FreeYes
Catalog No. 50-233-0603 Supplier Bellco Glass, Inc. Supplier No. 229148000
Tissue Culture Treated surface improves cell adhesionPlates are labeled with alpha-numeric markings for easy identification of individual wellsEach well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contaminationLids have a stacking feature for added stability when handling tall stacks of plates Pack DescriptionIndividual, 50/PackCase Quantity100SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPolystyreneTissue Culture TreatedYesBottom ShapeFlatCell Growth Area0.84cm²RNase/DNase FreeYes