Learn More
Brady™ 480/600V Breaker Blocker Kit
Catalog No. 19104178 Shop All Brady Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
19-104-178 1 Kit
1 options
Catalog No. 19-104-178 Supplier Brady™ Supplier No. 90891
Add to Cart
Add to Cart
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ 480/600V Breaker Blocker Kit is a great solution to safely lock out your oversized and irregularly shaped switches. Self-adhesive backed rails are permanently installed into electrical panels.
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Product Type Electrical Risk Lockouts
Includes (2) 4 in yellow mounting rails; (3) alcohol wipes; 7.6 in green blocking bar; 7.6 in red blocking bar
Quantity 1 Kit
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.