Learn More
Waters Corp Waters Corporation UV Wavelength Accuracy Standard, Uracil, 10 mL
Catalog No. 50818873 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This standard solution is designed for evaluating the wavelength accuracy of UV detectors. It contains 15.0 mg/L Uracil with >=98% purity in 100% HPLC Grade Methanol, supplied in a 10 mL volume.
  • Evaluates wavelength accuracy of UV detectors
  • Contains 15.0 mg/L Uracil, >=98% purity
  • Formulated in 100% HPLC Grade Methanol
  • 10 mL volume
  • Suitable for LC systems with Reversed Phase separation and UV detection
  • Room Temperature storage
  • Quantitative
  • 36 months shelf life
  • 0.05 tolerance
  • Reference type
  • System performance application
  • One unit per package
  • Hazardous material
Catalog No. 50-818-873 Supplier Waters Corp Supplier No. WAT042879
This standard solution is designed for evaluating the wavelength accuracy of UV detectors. It contains 15.0 mg/L Uracil with >=98% purity in 100% HPLC Grade Methanol, supplied in a 10 mL volume.
  • Evaluates wavelength accuracy of UV detectors
  • Contains 15.0 mg/L Uracil, >=98% purity
  • Formulated in 100% HPLC Grade Methanol
  • 10 mL volume
  • Suitable for LC systems with Reversed Phase separation and UV detection
  • Room Temperature storage
  • Quantitative
  • 36 months shelf life
  • 0.05 tolerance
  • Reference type
  • System performance application
  • One unit per package
  • Hazardous material