Learn More
ABclonal Technology 488 anti-Human CD225/IFITM1
SDP
Catalog No. 5000005821
Encompass_Preferred
Interferon-induced transmembrane (IFITM) proteins are a family of interferon induced antiviral proteins The family contains five members including IFITM1 IFITM2 and IFITM3 that belong to the CD225 superfamily The protein encoded by this gene restricts cellular entry by diverse viral pathogens such as influenza A virus Ebola virus and Sars-CoV-2
Catalog No. 50-000-05821 Supplier ABclonal Technology Supplier No. A22310200T
Interferon-induced transmembrane (IFITM) proteins are a family of interferon induced antiviral proteins The family contains five members including IFITM1 IFITM2 and IFITM3 that belong to the CD225 superfamily The protein encoded by this gene restricts cellular entry by diverse viral pathogens such as influenza A virus Ebola virus and Sars-CoV-2