Learn More
ABclonal Technology 488 anti-Human CD50ICAM-3
SDP
Catalog No. 5000011762
Encompass_Preferred
The protein encoded by this gene is a member of the intercellular adhesion molecule (ICAM) family All ICAM proteins are type I transmembrane glycoproteins contain 2-9 immunoglobulin-like C2-type domains and bind to the leukocyte adhesion LFA-1 protein This protein is constitutively and abundantly expressed by all leucocytes and may be the most important ligand for LFA-1 in the initiation of the immune response It functions not only as an adhesion molecule but also as a potent signalling molecule Alternative splicing results in multiple transcript variants encoding different isoforms
Catalog No. 50-000-11762 Supplier ABclonal Technology Supplier No. A25708100T
The protein encoded by this gene is a member of the intercellular adhesion molecule (ICAM) family All ICAM proteins are type I transmembrane glycoproteins contain 2-9 immunoglobulin-like C2-type domains and bind to the leukocyte adhesion LFA-1 protein This protein is constitutively and abundantly expressed by all leucocytes and may be the most important ligand for LFA-1 in the initiation of the immune response It functions not only as an adhesion molecule but also as a potent signalling molecule Alternative splicing results in multiple transcript variants encoding different isoforms