Learn More
ABclonal Technology 488 anti-Human/Monkey CD20
SDP
Catalog No. 5000007334
Encompass_Preferred
This gene encodes a member of the membrane-spanning 4A gene family Members of this nascent protein family are characterized by common structural features and similar intron exon splice boundaries and display unique expression patterns among hematopoietic cells and nonlymphoid tissues This gene encodes a B-lymphocyte surface molecule which plays a role in the development and differentiation of B-cells into plasma cells This family member is localized to 11q12 among a cluster of family members Alternative splicing of this gene results in two transcript variants which encode the same protein
Catalog No. 50-000-07334 Supplier ABclonal Technology Supplier No. A22152200T
This gene encodes a member of the membrane-spanning 4A gene family Members of this nascent protein family are characterized by common structural features and similar intron exon splice boundaries and display unique expression patterns among hematopoietic cells and nonlymphoid tissues This gene encodes a B-lymphocyte surface molecule which plays a role in the development and differentiation of B-cells into plasma cells This family member is localized to 11q12 among a cluster of family members Alternative splicing of this gene results in two transcript variants which encode the same protein