Learn More
ABclonal Technology 488 Conjugated AIF1/IBA1
SDP
Catalog No. 5000010332
Encompass_Preferred
Enables actin filament binding activity and calcium ion binding activity Involved in several processes including Rac protein signal transduction actin filament organization and ruffle assembly Acts upstream of or within actin filament bundle assembly Located in several cellular components including actin filament phagocytic cup and ruffle membrane Is expressed in adrenal cortex central nervous system embryo mesenchyme and retina Human ortholog(s) of this gene implicated in type 1 diabetes mellitus Orthologous to human AIF1 (allograft inflammatory factor 1)
Catalog No. 50-000-10332 Supplier ABclonal Technology Supplier No. A2603220UL
Enables actin filament binding activity and calcium ion binding activity Involved in several processes including Rac protein signal transduction actin filament organization and ruffle assembly Acts upstream of or within actin filament bundle assembly Located in several cellular components including actin filament phagocytic cup and ruffle membrane Is expressed in adrenal cortex central nervous system embryo mesenchyme and retina Human ortholog(s) of this gene implicated in type 1 diabetes mellitus Orthologous to human AIF1 (allograft inflammatory factor 1)