Learn More
ABclonal Technology 488 Rabbit anti-Human AGTR2
SDP
Catalog No. 5000008726
Encompass_Preferred
This gene encodes a G-protein coupled receptor 1 family member serving as a receptor for angiotensin II It is an integral membrane protein highly expressed in fetuses and neonates but less so in adults except in the brain adrenal medulla and atretic ovary This receptor mediates programmed cell death playing a role in developmental biology and disease pathophysiology Mutations are linked to X-linked cognitive disability SARS-CoV and SARS-CoV-2 infections downregulate ACE2 receptors triggering inflammatory lesions particularly in the lungs The angiotensin AT2 receptor is upregulated in bronchoalveolar lavage samples from COVID-19 patients suggesting its role in inflammation
Catalog No. 50-000-08726 Supplier ABclonal Technology Supplier No. A26592500T
This gene encodes a G-protein coupled receptor 1 family member serving as a receptor for angiotensin II It is an integral membrane protein highly expressed in fetuses and neonates but less so in adults except in the brain adrenal medulla and atretic ovary This receptor mediates programmed cell death playing a role in developmental biology and disease pathophysiology Mutations are linked to X-linked cognitive disability SARS-CoV and SARS-CoV-2 infections downregulate ACE2 receptors triggering inflammatory lesions particularly in the lungs The angiotensin AT2 receptor is upregulated in bronchoalveolar lavage samples from COVID-19 patients suggesting its role in inflammation