Learn More
ABclonal Technology 488 Rabbit anti-Human CaSR
SDP
Catalog No. 5000012061
Encompass_Preferred
The protein encoded by this gene is a plasma membrane G protein-coupled receptor that senses small changes in circulating calcium concentration The encoded protein couples this information to intracellular signaling pathways that modify parathyroid hormone secretion or renal cation handling and thus this protein plays an essential role in maintaining mineral ion homeostasis Mutations in this gene are a cause of familial hypocalciuric hypercalcemia neonatal severe hyperparathyroidism and autosomal dominant hypocalcemia
Catalog No. 50-000-12061 Supplier ABclonal Technology Supplier No. A26713200T
The protein encoded by this gene is a plasma membrane G protein-coupled receptor that senses small changes in circulating calcium concentration The encoded protein couples this information to intracellular signaling pathways that modify parathyroid hormone secretion or renal cation handling and thus this protein plays an essential role in maintaining mineral ion homeostasis Mutations in this gene are a cause of familial hypocalciuric hypercalcemia neonatal severe hyperparathyroidism and autosomal dominant hypocalcemia