Learn More
ABclonal Technology 488 Rabbit anti-Human CD172b
SDP
Catalog No. 5000013190
Encompass_Preferred
The protein encoded by this gene is a member of the signal-regulatory-protein (SIRP) family and also belongs to the immunoglobulin superfamily SIRP family members are receptor-type transmembrane glycoproteins known to be involved in the negative regulation of receptor tyrosine kinase-coupled signaling processes This protein was found to interact with TYROBP DAP12 a protein bearing immunoreceptor tyrosine-based activation motifs This protein was also reported to participate in the recruitment of tyrosine kinase SYK Multiple transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-13190 Supplier ABclonal Technology Supplier No. A25769100T
The protein encoded by this gene is a member of the signal-regulatory-protein (SIRP) family and also belongs to the immunoglobulin superfamily SIRP family members are receptor-type transmembrane glycoproteins known to be involved in the negative regulation of receptor tyrosine kinase-coupled signaling processes This protein was found to interact with TYROBP DAP12 a protein bearing immunoreceptor tyrosine-based activation motifs This protein was also reported to participate in the recruitment of tyrosine kinase SYK Multiple transcript variants encoding different isoforms have been found for this gene