Learn More
ABclonal Technology 488 Rabbit anti-Human CD1B
SDP
Supplier:  ABclonal Technology A26135500T
Encompass_Preferred
This gene encodes a member of the CD1 family of transmembrane glycoproteins which are structurally related to the major histocompatibility complex (MHC) proteins and form heterodimers with beta-2-microglobulin The CD1 proteins mediate the presentation of primarily lipid and glycolipid antigens of self or microbial origin to T cells The human genome contains five CD1 family genes organized in a cluster on chromosome 1 The CD1 family members are thought to differ in their cellular localization and specificity for particular lipid ligands The protein encoded by this gene localizes to late endosomes and lysosomes via a tyrosine-based motif in the cytoplasmic tail and requires vesicular acidification to bind lipid antigens
Catalog No. 50-000-09214