Learn More
ABclonal Technology 488 Rabbit anti-Human CD204
SDP
Catalog No. 5000011013
Encompass_Preferred
This gene encodes class A macrophage scavenger receptors which produce three isoforms (1 2 and 3) via alternative splicing These trimeric integral membrane glycoproteins are macrophage-specific and involved in various processes like atherosclerosis Alzheimer s disease and host defense Isoforms 1 and 2 mediate the endocytosis of modified low-density lipoproteins (LDLs) while isoform 3 despite having the necessary domain cannot internalize modified LDL due to altered intracellular processing and trapping within the endoplasmic reticulum Isoform 3 also inhibits the function of isoforms 1 and 2 when co-expressed suggesting a regulatory mechanism for scavenger receptor activity in macrophages
Catalog No. 50-000-11013 Supplier ABclonal Technology Supplier No. A25612500T
This gene encodes class A macrophage scavenger receptors which produce three isoforms (1 2 and 3) via alternative splicing These trimeric integral membrane glycoproteins are macrophage-specific and involved in various processes like atherosclerosis Alzheimer s disease and host defense Isoforms 1 and 2 mediate the endocytosis of modified low-density lipoproteins (LDLs) while isoform 3 despite having the necessary domain cannot internalize modified LDL due to altered intracellular processing and trapping within the endoplasmic reticulum Isoform 3 also inhibits the function of isoforms 1 and 2 when co-expressed suggesting a regulatory mechanism for scavenger receptor activity in macrophages