Learn More
ABclonal Technology 488 Rabbit anti-Human CD30
Supplier: ABclonal Technology A22214200T
The protein encoded by this gene is a member of the TNF-receptor superfamily This receptor is expressed by activated but not by resting T and B cells TRAF2 and TRAF5 can interact with this receptor and mediate the signal transduction that leads to the activation of NF-kappaB This receptor is a positive regulator of apoptosis and also has been shown to limit the proliferative potential of autoreactive CD8 effector T cells and protect the body against autoimmunity Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported