Learn More
ABclonal Technology 488 Rabbit anti-Human CD48
SDP
Catalog No. 5000006638
Encompass_Preferred
This gene encodes a member of the CD2 subfamily of immunoglobulin-like receptors which includes SLAM (signaling lymphocyte activation molecules) proteins The encoded protein is found on the surface of lymphocytes and other immune cells dendritic cells and endothelial cells and participates in activation and differentiation pathways in these cells The encoded protein does not have a transmembrane domain however but is held at the cell surface by a GPI anchor via a C-terminal domain which maybe cleaved to yield a soluble form of the receptor Multiple transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-06638 Supplier ABclonal Technology Supplier No. A21937100T
This gene encodes a member of the CD2 subfamily of immunoglobulin-like receptors which includes SLAM (signaling lymphocyte activation molecules) proteins The encoded protein is found on the surface of lymphocytes and other immune cells dendritic cells and endothelial cells and participates in activation and differentiation pathways in these cells The encoded protein does not have a transmembrane domain however but is held at the cell surface by a GPI anchor via a C-terminal domain which maybe cleaved to yield a soluble form of the receptor Multiple transcript variants encoding different isoforms have been found for this gene