Learn More
ABclonal Technology 488 Rabbit anti-Human CD51
SDP
Catalog No. 5000007455
Encompass_Preferred
The product of this gene belongs to the integrin alpha chain family Integrins are heterodimeric integral membrane proteins composed of an alpha subunit and a beta subunit that function in cell surface adhesion and signaling The encoded preproprotein is proteolytically processed to generate light and heavy chains that comprise the alpha V subunit This subunit associates with beta 1 beta 3 beta 5 beta 6 and beta 8 subunits The heterodimer consisting of alpha V and beta 3 subunits is also known as the vitronectin receptor This integrin may regulate angiogenesis and cancer progression Alternative splicing results in multiple transcript variants Note that the integrin alpha 5 and integrin alpha V subunits are encoded by distinct genes
Catalog No. 50-000-07455 Supplier ABclonal Technology Supplier No. A22298100T
The product of this gene belongs to the integrin alpha chain family Integrins are heterodimeric integral membrane proteins composed of an alpha subunit and a beta subunit that function in cell surface adhesion and signaling The encoded preproprotein is proteolytically processed to generate light and heavy chains that comprise the alpha V subunit This subunit associates with beta 1 beta 3 beta 5 beta 6 and beta 8 subunits The heterodimer consisting of alpha V and beta 3 subunits is also known as the vitronectin receptor This integrin may regulate angiogenesis and cancer progression Alternative splicing results in multiple transcript variants Note that the integrin alpha 5 and integrin alpha V subunits are encoded by distinct genes