Learn More
ABclonal Technology 488 Rabbit anti-Human CD70
Supplier: ABclonal Technology A22302200T
The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family This cytokine is a ligand for TNFRSF27 CD27 It is a surface antigen on activated but not on resting T and B lymphocytes It induces proliferation of costimulated T cells enhances the generation of cytolytic T cells and contributes to T cell activation This cytokine is also reported to play a role in regulating B-cell activation cytotoxic function of natural killer cells and immunoglobulin sythesis