Learn More
ABclonal Technology 488 Rabbit anti-Human CD75
Supplier: ABclonal Technology A24683200T
This gene encodes a member of glycosyltransferase family 29 The encoded protein is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates The protein which is normally found in the Golgi but can be proteolytically processed to a soluble form is involved in the generation of the cell-surface carbohydrate determinants and differentiation antigens HB-6 CD75 and CD76 This gene has been incorrectly referred to as CD75 Alternative splicing results in multiple transcript variants