Learn More
ABclonal Technology 488 Rabbit anti-Human CD8b
SDP
Catalog No. 5000012354
Encompass_Preferred
The CD8 antigen is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediates efficient cell-cell interactions within the immune system The CD8 antigen acting as a coreceptor and the T-cell receptor on the T lymphocyte recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules The functional coreceptor is either a homodimer composed of two alpha chains or a heterodimer composed of one alpha and one beta chain Both alpha and beta chains share significant homology to immunoglobulin variable light chains This gene encodes the CD8 beta chain isoforms Multiple alternatively spliced transcript variants encoding distinct membrane associated or secreted isoforms have been described A pseudogene also located on chromosome 2 has been identified
Catalog No. 50-000-12354 Supplier ABclonal Technology Supplier No. A24986100T
The CD8 antigen is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediates efficient cell-cell interactions within the immune system The CD8 antigen acting as a coreceptor and the T-cell receptor on the T lymphocyte recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules The functional coreceptor is either a homodimer composed of two alpha chains or a heterodimer composed of one alpha and one beta chain Both alpha and beta chains share significant homology to immunoglobulin variable light chains This gene encodes the CD8 beta chain isoforms Multiple alternatively spliced transcript variants encoding distinct membrane associated or secreted isoforms have been described A pseudogene also located on chromosome 2 has been identified