Learn More
ABclonal Technology 488 Rabbit anti-Human CD94
SDP
Catalog No. 5000005507
Encompass_Preferred
Natural killer (NK) cells are a distinct lineage of lymphocytes that mediate cytotoxic activity and secrete cytokines upon immune stimulation Several genes of the C-type lectin superfamily including members of the NKG2 family are expressed by NK cells and may be involved in the regulation of NK cell function KLRD1 (CD94) is an antigen preferentially expressed on NK cells and is classified as a type II membrane protein because it has an external C terminus Three transcript variants encoding two different isoforms have been found for this gene
Catalog No. 50-000-05507 Supplier ABclonal Technology Supplier No. A23804100T
Natural killer (NK) cells are a distinct lineage of lymphocytes that mediate cytotoxic activity and secrete cytokines upon immune stimulation Several genes of the C-type lectin superfamily including members of the NKG2 family are expressed by NK cells and may be involved in the regulation of NK cell function KLRD1 (CD94) is an antigen preferentially expressed on NK cells and is classified as a type II membrane protein because it has an external C terminus Three transcript variants encoding two different isoforms have been found for this gene