Learn More
ABclonal Technology 488 Rabbit anti-Human CDA
SDP
Catalog No. 5000009625
Encompass_Preferred
This gene encodes an enzyme involved in pyrimidine salvaging The encoded protein forms a homotetramer that catalyzes the irreversible hydrolytic deamination of cytidine and deoxycytidine to uridine and deoxyuridine respectively It is one of several deaminases responsible for maintaining the cellular pyrimidine pool Mutations in this gene are associated with decreased sensitivity to the cytosine nucleoside analogue cytosine arabinoside used in the treatment of certain childhood leukemias
Catalog No. 50-000-09625 Supplier ABclonal Technology Supplier No. A24685200T
This gene encodes an enzyme involved in pyrimidine salvaging The encoded protein forms a homotetramer that catalyzes the irreversible hydrolytic deamination of cytidine and deoxycytidine to uridine and deoxyuridine respectively It is one of several deaminases responsible for maintaining the cellular pyrimidine pool Mutations in this gene are associated with decreased sensitivity to the cytosine nucleoside analogue cytosine arabinoside used in the treatment of certain childhood leukemias