Learn More
ABclonal Technology 488 Rabbit anti-Human EGFR
SDP
Supplier:  ABclonal Technology A24265100T
Encompass_Preferred
The protein encoded by this gene is a transmembrane glycoprotein that is a member of the protein kinase superfamily This protein is a receptor for members of the epidermal growth factor family EGFR is a cell surface protein that binds to epidermal growth factor Binding of the protein to a ligand induces receptor dimerization and tyrosine autophosphorylation and leads to cell proliferation Mutations in this gene are associated with lung cancer
Catalog No. 50-000-09374