Learn More
ABclonal Technology 488 Rabbit anti-Human FATP1
SDP
Supplier:  ABclonal Technology A25570200T
Encompass_Preferred
Enables biotin transmembrane transporter activity efflux transmembrane transporter activity and long-chain fatty acid transporter activity Involved in several processes including carboxylic acid transmembrane transport glycerophospholipid biosynthetic process and lipid transport across blood-brain barrier Located in membrane Part of plasma membrane
Catalog No. 50-000-11335