Learn More
ABclonal Technology 488 Rabbit anti-Human IL-2
SDP
Catalog No. 5000009649
Encompass_Preferred
This gene is a member of the interleukin 2 (IL2) cytokine subfamily which includes IL4 IL7 IL9 IL15 IL21 erythropoietin and thrombopoietin The protein encoded by this gene is a secreted cytokine produced by activated CD4 and CD8 T lymphocytes that is important for the proliferation of T and B lymphocytes The receptor of this cytokine (IL2R) is a heterotrimeric protein complex whose gamma chain is also shared by IL4 and IL7 The expression of this gene in mature thymocytes is monoallelic which represents an unusual regulatory mode for controlling the precise expression of a single gene The targeted disruption of a similar gene in mice leads to ulcerative colitis-like disease which suggests an essential role of this gene in the immune response to antigenic stimuli
Catalog No. 50-000-09649 Supplier ABclonal Technology Supplier No. A27426500T
This gene is a member of the interleukin 2 (IL2) cytokine subfamily which includes IL4 IL7 IL9 IL15 IL21 erythropoietin and thrombopoietin The protein encoded by this gene is a secreted cytokine produced by activated CD4 and CD8 T lymphocytes that is important for the proliferation of T and B lymphocytes The receptor of this cytokine (IL2R) is a heterotrimeric protein complex whose gamma chain is also shared by IL4 and IL7 The expression of this gene in mature thymocytes is monoallelic which represents an unusual regulatory mode for controlling the precise expression of a single gene The targeted disruption of a similar gene in mice leads to ulcerative colitis-like disease which suggests an essential role of this gene in the immune response to antigenic stimuli