Learn More
ABclonal Technology 488 Rabbit anti-Human KLRG1
SDP
Catalog No. 5000012646
Encompass_Preferred
Natural killer (NK) cells are lymphocytes that can mediate lysis of certain tumor cells and virus-infected cells without previous activation They can also regulate specific humoral and cell-mediated immunity The protein encoded by this gene belongs to t
Catalog No. 50-000-12646 Supplier ABclonal Technology Supplier No. A26140PM200T
Natural killer (NK) cells are lymphocytes that can mediate lysis of certain tumor cells and virus-infected cells without previous activation They can also regulate specific humoral and cell-mediated immunity The protein encoded by this gene belongs to t