Learn More
ABclonal Technology 488 Rabbit anti-Human LAMP2
SDP
Catalog No. 5000005172
Encompass_Preferred
The protein encoded by this gene is a member of a family of membrane glycoproteins This glycoprotein provides selectins with carbohydrate ligands It may play a role in tumor cell metastasis It may also function in the protection maintenance and adhesion of the lysosome Alternative splicing of this gene results in multiple transcript variants encoding distinct proteins
Catalog No. 50-000-05172 Supplier ABclonal Technology Supplier No. A22216500T
The protein encoded by this gene is a member of a family of membrane glycoproteins This glycoprotein provides selectins with carbohydrate ligands It may play a role in tumor cell metastasis It may also function in the protection maintenance and adhesion of the lysosome Alternative splicing of this gene results in multiple transcript variants encoding distinct proteins