Learn More
ABclonal Technology 488 Rabbit anti-Human MRC2
SDP
Supplier:  ABclonal Technology A26562500T
Encompass_Preferred
This gene encodes a member of the mannose receptor family of proteins that contain a fibronectin type II domain and multiple C-type lectin-like domains The encoded protein plays a role in extracellular matrix remodeling by mediating the internalization and lysosomal degradation of collagen ligands Expression of this gene may play a role in the tumorigenesis and metastasis of several malignancies including breast cancer gliomas and metastatic bone disease
Catalog No. 50-000-12232