Learn More
ABclonal Technology 488 Rabbit anti-Human SIGLEC9
SDP
Catalog No. 5000010764
Encompass_Preferred
Predicted to enable monosaccharide binding activity and sialic acid binding activity Predicted to be involved in cell adhesion Predicted to act upstream of or within negative regulation of inflammatory response and negative regulation of phagocytosis engulfment Predicted to be located in external side of plasma membrane Predicted to be active in plasma membrane
Catalog No. 50-000-10764 Supplier ABclonal Technology Supplier No. A25868200T
Predicted to enable monosaccharide binding activity and sialic acid binding activity Predicted to be involved in cell adhesion Predicted to act upstream of or within negative regulation of inflammatory response and negative regulation of phagocytosis engulfment Predicted to be located in external side of plasma membrane Predicted to be active in plasma membrane