Learn More
ABclonal Technology 488 Rabbit anti-Human Syk
SDP
Catalog No. 5000013184
Encompass_Preferred
This gene encodes a member of the family of non-receptor type Tyr protein kinases This protein is widely expressed in hematopoietic cells and is involved in coupling activated immunoreceptors to downstream signaling events that mediate diverse cellular responses including proliferation differentiation and phagocytosis It is thought to be a modulator of epithelial cell growth and a potential tumour suppressor in human breast carcinomas Alternatively spliced transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-13184 Supplier ABclonal Technology Supplier No. A25738200T
This gene encodes a member of the family of non-receptor type Tyr protein kinases This protein is widely expressed in hematopoietic cells and is involved in coupling activated immunoreceptors to downstream signaling events that mediate diverse cellular responses including proliferation differentiation and phagocytosis It is thought to be a modulator of epithelial cell growth and a potential tumour suppressor in human breast carcinomas Alternatively spliced transcript variants encoding different isoforms have been found for this gene