Learn More
ABclonal Technology 488 Rabbit anti-Human TLR1
SDP
Catalog No. 5000009991
Encompass_Preferred
The protein encoded by this gene is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity TLRs are highly conserved from Drosophila to humans and share structural and functional similarities They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents and mediate the production of cytokines necessary for the development of effective immunity The various TLRs exhibit different patterns of expression This gene is ubiquitously expressed and at higher levels than other TLR genes Different length transcripts presumably resulting from use of alternative polyadenylation site and or from alternative splicing have been noted for this gene
Catalog No. 50-000-09991 Supplier ABclonal Technology Supplier No. A26257100T
The protein encoded by this gene is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity TLRs are highly conserved from Drosophila to humans and share structural and functional similarities They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents and mediate the production of cytokines necessary for the development of effective immunity The various TLRs exhibit different patterns of expression This gene is ubiquitously expressed and at higher levels than other TLR genes Different length transcripts presumably resulting from use of alternative polyadenylation site and or from alternative splicing have been noted for this gene