Learn More
ABclonal Technology 488 Rabbit anti-Human TREM1
SDP
Supplier:  ABclonal Technology A26872500T
Encompass_Preferred
This gene encodes a receptor belonging to the Ig superfamily that is expressed on myeloid cells This protein amplifies neutrophil and monocyte-mediated inflammatory responses triggered by bacterial and fungal infections by stimulating release of pro-inflammatory chemokines and cytokines as well as increased surface expression of cell activation markers Alternatively spliced transcript variants encoding different isoforms have been noted for this gene
Catalog No. 50-000-08527