Learn More
ABclonal Technology 488 Rabbit anti-Human TROP-2
Supplier: ABclonal Technology A22577200T
This intronless gene encodes a carcinoma-associated antigen This antigen is a cell surface receptor that transduces calcium signals Mutations of this gene have been associated with gelatinous drop-like corneal dystrophy