Learn More
ABclonal Technology 488 Rabbit anti-Human TROP-2
SDP
Catalog No. 5000009706
Encompass_Preferred
This intronless gene encodes a carcinoma-associated antigen This antigen is a cell surface receptor that transduces calcium signals Mutations of this gene have been associated with gelatinous drop-like corneal dystrophy
Catalog No. 50-000-09706 Supplier ABclonal Technology Supplier No. A24360100T
This intronless gene encodes a carcinoma-associated antigen This antigen is a cell surface receptor that transduces calcium signals Mutations of this gene have been associated with gelatinous drop-like corneal dystrophy