Learn More
ABclonal Technology 488 Rabbit anti-Human ULBP2
SDP
Catalog No. 5000005145
Encompass_Preferred
This gene encodes a major histocompatibility complex (MHC) class I-related molecule that binds to the NKG2D receptor on natural killer (NK) cells to trigger release of multiple cytokines and chemokines that in turn contribute to the recruitment and activation of NK cells The encoded protein undergoes further processing to generate the mature protein that is either anchored to membrane via a glycosylphosphatidylinositol moiety or secreted Many malignant cells secrete the encoded protein to evade immunosurveillance by NK cells This gene is located in a cluster of multiple MHC class I-related genes on chromosome 6
Catalog No. 50-000-05145 Supplier ABclonal Technology Supplier No. A22694200T
This gene encodes a major histocompatibility complex (MHC) class I-related molecule that binds to the NKG2D receptor on natural killer (NK) cells to trigger release of multiple cytokines and chemokines that in turn contribute to the recruitment and activation of NK cells The encoded protein undergoes further processing to generate the mature protein that is either anchored to membrane via a glycosylphosphatidylinositol moiety or secreted Many malignant cells secrete the encoded protein to evade immunosurveillance by NK cells This gene is located in a cluster of multiple MHC class I-related genes on chromosome 6