Learn More
Uline Cool Shield Thermal Bubble Roll - 48"""" x 125'
Catalog No. NC2597707
Encompass
Clear plastic bubble sheeting for providing a protective barrier or cover, 3/16"""" bubble cushions, metalized polyester on both sides, cannot be used with dry ice
Catalog No. NC2597707 Supplier Uline Supplier No. S9911
Clear plastic bubble sheeting for providing a protective barrier or cover, 3/16"""" bubble cushions, metalized polyester on both sides, cannot be used with dry ice