Learn More
NEST SCIENTIFIC INC 48 Well Cell Culture Plate, Flat, Non-Treated, sterile 50/cs
Catalog No. NC3454543
Encompass_Preferred
Nest Scientific cell culture plates are molded from crystal clear USP grade VI virgin polystyrene. This part number comes indovidually wrapped in Tyvek/plastic blister packages. Plates are untreated to support suspended lines. Molded in an ISO 7 cleanroom, sterile (SAL 1 X 10^-6) and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. cGMP compliant (21 CFR part 820/821). Extensive validation dossier available upon request.
Catalog No. NC3454543 Supplier NEST SCIENTIFIC INC Supplier No. 748011
Nest Scientific cell culture plates are molded from crystal clear USP grade VI virgin polystyrene. This part number comes indovidually wrapped in Tyvek/plastic blister packages. Plates are untreated to support suspended lines. Molded in an ISO 7 cleanroom, sterile (SAL 1 X 10^-6) and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. cGMP compliant (21 CFR part 820/821). Extensive validation dossier available upon request.