Learn More
Uline Additional Chrome Wire Shelves - 48 x 24", 2/pack
Catalog No. NC0770837
Encompass
48 x 24" chrome wire shelves, chrome-plated steel, snap-on plastic shelf supports, 800lbs capacity (based on evenly distributed loads)
Catalog No. NC0770837 Supplier Uline Supplier No. H3187C
48 x 24" chrome wire shelves, chrome-plated steel, snap-on plastic shelf supports, 800lbs capacity (based on evenly distributed loads)