Learn More
Felix Storch Inc ACCUCOLD 49 CU/FT DOUB/DOOR REFRIG
SDP
Catalog No. NC2681232
Encompass_Preferred
The SCRR492 is a two-door commercial all-refrigerator with a complete stainless steel cabinet. The doors are self-closing to help protect stored contents in busy commercial settings where customers may not take the time to make sure the doors are closed. They will remain open when pushed to 90º, making it easier to load and unload the unit. The sealed back design helps to simplify cleanup. This unit features a factory-installed lock located towards the top of the door for convenient security. We include a set of locking casters for optional installation: https://www.accucold.com/product/SCRR492
Catalog No. NC2681232 Supplier Felix Storch Inc Supplier No. SCRR492
Add to Cart
Add to Cart
Request a Quote
The SCRR492 is a two-door commercial all-refrigerator with a complete stainless steel cabinet. The doors are self-closing to help protect stored contents in busy commercial settings where customers may not take the time to make sure the doors are closed. They will remain open when pushed to 90º, making it easier to load and unload the unit. The sealed back design helps to simplify cleanup. This unit features a factory-installed lock located towards the top of the door for convenient security. We include a set of locking casters for optional installation: https://www.accucold.com/product/SCRR492