Learn More
Andwin Scientific 49-Place Fiberboard Cell Divider
SDP
Catalog No. NC1729223
Encompass_Preferred
VWR Fiberboard Dividers 49 cell. Dimensions: 5 x 5 x 1 inches. Can be utilized in any 5 x 5 inches mechanical or liquid nitrogen cryogenic freezer box.
Catalog No. NC1729223 Supplier Andwin Scientific Supplier No. 82007150
VWR Fiberboard Dividers 49 cell. Dimensions: 5 x 5 x 1 inches. Can be utilized in any 5 x 5 inches mechanical or liquid nitrogen cryogenic freezer box.