Learn More
Abcam 490/515 nm E x tracellular Vesicle Membrane Staining Kit, 100 TestsS
Catalog No. 502699012
Encompass_Preferred
Labeling in < 4 hrs with 30 secs hands-on time using 490/515 nm Extracellular Vesicle Membrane Staining Kit ab314686. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-269-9012 Supplier Abcam Supplier No. AB314686100TESTS
Labeling in < 4 hrs with 30 secs hands-on time using 490/515 nm Extracellular Vesicle Membrane Staining Kit ab314686. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement