Learn More
Aviva Systems Biology Corp SHTN1 Antibody - N-terminal region (ARP57205_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57205_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against 4930506M07Rik. It was validated on Western Blot. . Alternate names: Kiaa1598, Shootin1, mKIAA1598ApplicationWB. Species reactivity: Horse, Human, Mouse, Pig, Rat, Yeast
Catalog No. 50-286-279