Learn More
Brady™ Universal Valve Lockout Base Clamping Units
Catalog No. p-4932392 Shop All Brady Corporation Products
Change view
Click to view available options
Product Type:
Large Base Clamping Unit
Small Base Clamping unit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-000-281 Small Base Clamping unit
19-000-280 Large Base Clamping Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-000-281 Supplier Brady™ Supplier No. 50924
Only null left
Add to Cart
Add to Cart
Used to lock out large levers, T-handles and hard-to-secure mechanical devices
  • Base unit only (cable and blocking arm sold separately)
  • Rugged, easy to use and lightweight
  • Made of Industrial-Grade Steel and Nylon
  • Open-ended clamp fits over closed rings and wide handles
  • Available in small and large sizes
Specifications
Product Type Small Base Clamping unit
Color Red
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.