Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Arrow Tapes

Catalog No. 19000343 Shop All Brady Corporation Products
Change view
Click to view available options
Display Text:
270 Arrows
720 Arrows
Dimensions (L x W):
30 yd. x 2 in.
30 yd. x 4 in.
Color:
Blue, White
Red, White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text Dimensions (L x W) Color
19-000-343 720 Arrows 30 yd. x 2 in. Red, White
19-000-344 720 Arrows 30 yd. x 2 in. Blue, White
19-000-342 270 Arrows 30 yd. x 4 in. Blue, White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-000-343 Supplier Brady™ Supplier No. 91422
Add to Cart
Add to Cart

Directional tapes to enhance safety, improve operating effciency and provide 360 degree visibility

  • Made of B-946 pressure sensitive vinyl withstands elements indoors or out
  • Supplied in liner-mounted rolled form

Specifications

Adhesive Type Acrylic
Color Red, White
Finish Gloss
Material Vinyl
Display Text 720 Arrows
For Use With (Application) Pipe and Valve Marking
Quantity 1 Ea.
Dimensions (L x W) 30 yd. x 2 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.