Learn More
Brady™ Socks
Catalog No. 19000176 Shop All Brady Corporation Products
Change view
Click to view available options
Packaging Quantity:
12/Cs.
30/Cs.
6/Cs.
Length (English):
4 ft.
8 ft.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity Length (English)
19-000-176 6/Cs. 8 ft.
19-000-174 12/Cs. 4 ft.
19-000-175 30/Cs. 4 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-000-176 Supplier Brady™ Supplier No. AW806
  • $244.00
    $167.65 / Case of 6
    Save $76.35
    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Dependable, flexible tubes that contain and absorb liquids
  • Made of all-purpose polypropylene SOC
  • Molds around corners and conforms to uneven surfaces
  • Wringable and can be incinerated
  • Used as a working tool for all major industries
  • Available in variety of types and sizes
Specifications
Material Polypropylene
Type Allwik Socks
For Use With (Application) Industrial applications
Packaging Quantity 6/Cs.
Length (English) 8 ft.
Diameter (English) 3 in.