Learn More
Brady™ Oil Only Pillows
Catalog No. 19000168 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
28 gal./Bale
Catalog No. Capacity (English)
19-000-168 28 gal./Bale
1 options
Catalog No. 19-000-168 Supplier Brady™ Supplier No. OIL1818
  • $196.00
    $136.65 / Case of 16
    Save $59.35
    30% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
For absorbing oil and petroleum based spills and leaking fluids in tight spaces
  • Feature white spunbond outer covering filled with white polypropylene
  • For use in waste troughs, coolant tanks, fluid reservoirs and sump wells
  • Specifications
    Capacity (English) 28 gal./Bale
    Material Polypropylene
    Type Oil Only Pillow
    For Use With (Application) Overspray leaks and drips, drums - liquid storage, drains
    Packaging Quantity 16/Cs.